siRNA Details
| Gene Symbol: Atp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139106.1 | NP_620806.1 | 2 | 335 | ||
| XM_006240845.2 | XP_006240907.1 | 3 | 850 | |||
| GenBank mRNA | FQ214102.1 | - | 543 | |||
| FQ223899.1 | - | 2 | 383 | |||
| U00926.1 | - | 2 | 335 | |||

Atp5d
Genomic Map