siRNA Details
| Gene Symbol: HTR1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000865.2 | NP_000856.1 | 2 | 1974 | ||
| XM_011535789.2 | XP_011534091.1 | 2 | 2422 | |||
| GenBank mRNA | BC069751.1 | - | 2 | 1553 | 7262611 | |
| M91467.1 | - | 3 | 1837 | |||
More Information
Set Membership: |
|

HTR1E
Genomic Map