siRNA Details
| Gene Symbol: Klrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139187.2 | NP_631926.1 | 6 | 526 | ||
| GenBank mRNA | AF416564.2 | - | 6 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrh1| Gene Symbol: Klrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139187.2 | NP_631926.1 | 6 | 526 | ||
| GenBank mRNA | AF416564.2 | - | 6 | 526 | ||