siRNA Details
| Gene Symbol: Ly6i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139257.1 | NP_640350.1 | 4 | 807 | ||
| XM_006241765.2 | XP_006241827.1 | 794 | ||||
| XM_006241766.2 | XP_006241828.1 | 5 | 934 | |||
| XM_008765545.1 | XP_008763767.1 | 5 | 921 | |||
| GenBank mRNA | M30689.1 | - | 4 | 807 | ||

Ly6i
Genomic Map