siRNA Details
| Gene Symbol: Hsd3b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139329.1 | NP_647545.1 | 7 | 1619 | ||
| XM_006230212.2 | XP_006230274.1 | 7 | 2098 | |||
| GenBank mRNA | AB000199.1 | - | 7 | 1619 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd3b7| Gene Symbol: Hsd3b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139329.1 | NP_647545.1 | 7 | 1619 | ||
| XM_006230212.2 | XP_006230274.1 | 7 | 2098 | |||
| GenBank mRNA | AB000199.1 | - | 7 | 1619 | ||