siRNA Details
| Gene Symbol: Acsm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144748.1 | NP_653349.1 | 7, 8 | 1077 | ||
| XM_008759677.1 | XP_008757899.1 | 7, 8 | 1062 | |||
| XM_008759678.1 | XP_008757900.1 | 7, 8 | 1099 | |||
| GenBank mRNA | AF062389.1 | - | 7, 8 | 1077 | ||
| BC078721.1 | - | 7, 8 | 1054 | 7110246 | ||

Acsm2a
Genomic Map