siRNA Details
| Gene Symbol: Slc15a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144758.1 | NP_653359.1 | 1, 2 | 572 | ||
| GenBank mRNA | AB000280.1 | - | 1, 2 | 572 | ||
| BC087709.1 | - | 1, 2 | 571 | 7315369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc15a4| Gene Symbol: Slc15a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144758.1 | NP_653359.1 | 1, 2 | 572 | ||
| GenBank mRNA | AB000280.1 | - | 1, 2 | 572 | ||
| BC087709.1 | - | 1, 2 | 571 | 7315369 | ||