siRNA Details
| Gene Symbol: Tor1aip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145092.2 | NP_659560.2 | 10 | 2269 | ||
| XM_006250027.2 | XP_006250089.1 | 10 | 1814 | |||
| XM_006250028.2 | XP_006250090.1 | 10 | 2400 | |||
| GenBank mRNA | BC086987.1 | - | 10 | 2269 | 7129718 | |
| FQ234456.1 | - | 1 | 938 | |||
| U19614.1 | - | 10 | 1908 | |||
| U20286.1 | - | 10 | 1726 | |||

Tor1aip1
Genomic Map