siRNA Details
Gene Symbol: Cyp4x1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145675.1 | NP_663708.1 | 2, 3 | 319 | ||
GenBank mRNA | AF439343.1 | - | 2, 3 | 319 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
