siRNA Details
| Gene Symbol: Cdk5rap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145721.2 | NP_663773.1 | 1 | 227 | ||
| XM_006235306.1 | XP_006235368.1 | 2 | 542 | |||
| XM_006235307.2 | XP_006235369.1 | 3 | 433 | |||
| XM_008762302.1 | XP_008760524.1 | 3 | 436 | |||
| GenBank mRNA | AF177477.1 | - | 236 | |||
| BC066666.1 | - | 1 | 227 | 5623565 | ||

Cdk5rap1
Genomic Map