siRNA Details
Gene Symbol: Zfp394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145724.1 | NP_663776.1 | 3 | 1157 | ||
GenBank mRNA | AF052042.1 | - | 3 | 1157 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145724.1 | NP_663776.1 | 3 | 1157 | ||
GenBank mRNA | AF052042.1 | - | 3 | 1157 |