siRNA Details
| Gene Symbol: Drgx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145767.1 | NP_665710.1 | 3, 4 | 402 | ||
| GenBank mRNA | U29174.1 | - | 3, 4 | 402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Drgx| Gene Symbol: Drgx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145767.1 | NP_665710.1 | 3, 4 | 402 | ||
| GenBank mRNA | U29174.1 | - | 3, 4 | 402 | ||