siRNA Details
Gene Symbol: Mamdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145768.1 | NP_665711.1 | 2 | 168 | ||
GenBank mRNA | L37380.1 | - | 2 | 168 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mamdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145768.1 | NP_665711.1 | 2 | 168 | ||
GenBank mRNA | L37380.1 | - | 2 | 168 |