siRNA Details
Gene Symbol: LOC257643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147137.3 | NP_671478.2 | 2 | 295 | ||
GenBank mRNA | AF442205.1 | - | 2 | 325 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC257643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147137.3 | NP_671478.2 | 2 | 295 | ||
GenBank mRNA | AF442205.1 | - | 2 | 325 |