siRNA Details
| Gene Symbol: RASSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164746.1 | NP_001158218.1 | 2 | 630 | ||
| NM_001164747.1 | NP_001158219.1 | 3 | 375 | |||
| NM_001164748.1 | NP_001158220.1 | 2 | 309 | |||
| NM_007211.4 | NP_009142.2 | 3 | 403 | |||
| XM_006719027.2 | XP_006719090.1 | 2 | 177 | |||
| XM_006719028.1 | XP_006719091.1 | 2 | 581 | |||
| XM_017018746.1 | XP_016874235.1 | 2 | 217 | |||
| GenBank mRNA | AB093206.1 | - | 2 | 282 | ||
| AK290055.1 | - | 3 | 375 | |||
| AK292333.1 | - | 2 | 624 | |||
| AY665468.1 | - | 2 | 232 | |||
| BC030021.1 | - | 3 | 405 | 5266519 | ||
| CR456905.1 | - | 1 | 83 | |||
| DB044325.1 | - | 2 | 270 | |||
| U82396.1 | - | 1 | 205 | |||

RASSF8
Genomic Map