siRNA Details
| Gene Symbol: Aurka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153296.2 | NP_695208.2 | 2 | 221 | ||
| XM_006235675.2 | XP_006235737.1 | 3 | 300 | |||
| XM_006235676.2 | XP_006235738.1 | 3 | 317 | |||
| GenBank mRNA | AF537333.1 | - | 2 | 209 | ||
| BC104676.1 | - | 2 | 221 | 7453115 | ||
| FQ227101.1 | - | 3 | 307 | |||

Aurka
Genomic Map