siRNA Details
Gene Symbol: Slc35e4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153316.2 | NP_695228.2 | 1 | 723 | ||
GenBank mRNA | AF182714.1 | - | 1 | 730 | ||
BC085693.1 | - | 1 | 723 | 7191218 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc35e4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153316.2 | NP_695228.2 | 1 | 723 | ||
GenBank mRNA | AF182714.1 | - | 1 | 730 | ||
BC085693.1 | - | 1 | 723 | 7191218 |