siRNA Details
| Gene Symbol: Npas4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153626.1 | NP_705890.1 | 5 | 776 | ||
| XM_006230697.2 | XP_006230759.1 | 7 | 2933 | |||
| GenBank mRNA | AB050103.1 | - | 5 | 776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npas4| Gene Symbol: Npas4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153626.1 | NP_705890.1 | 5 | 776 | ||
| XM_006230697.2 | XP_006230759.1 | 7 | 2933 | |||
| GenBank mRNA | AB050103.1 | - | 5 | 776 | ||