siRNA Details
| Gene Symbol: Gpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153727.1 | NP_714949.1 | 1 | 439 | ||
| XM_006239006.2 | XP_006239068.1 | 2 | 578 | |||
| GenBank mRNA | AJ427482.1 | - | 1 | 439 | ||
More Information
Set Membership: |
|

Gpr3
Genomic Map