siRNA Details
Gene Symbol: Zfp597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153732.2 | NP_714954.2 | 3 | 1473 | ||
GenBank mRNA | AF277900.2 | - | 1349 | |||
BC078709.1 | - | 3 | 1473 | 7113822 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153732.2 | NP_714954.2 | 3 | 1473 | ||
GenBank mRNA | AF277900.2 | - | 1349 | |||
BC078709.1 | - | 3 | 1473 | 7113822 |