siRNA Details
Gene Symbol: Duox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153739.1 | NP_714961.1 | 33 | 4894 | ||
XM_006234829.2 | XP_006234891.1 | 34 | 4993 | |||
GenBank mRNA | AF542180.1 | - | 33 | 4894 |
