siRNA Details
| Gene Symbol: Sclt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153740.1 | NP_714962.1 | 6 | 360 | ||
| XM_006232296.2 | XP_006232358.1 | 6 | 822 | |||
| GenBank mRNA | AF421190.1 | - | 6 | 360 | ||
| AF421191.1 | - | 6 | 355 | |||
| AF427094.1 | - | 6 | 360 | |||
| AF427095.1 | - | 5 | 309 | |||

Sclt1
Genomic Map