siRNA Details
Gene Symbol: Ppp1r14d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172011.1 | NP_742008.1 | 4 | 585 | ||
GenBank mRNA | AY122322.1 | - | 4 | 585 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppp1r14d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172011.1 | NP_742008.1 | 4 | 585 | ||
GenBank mRNA | AY122322.1 | - | 4 | 585 |