siRNA Details
| Gene Symbol: Dus1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172018.1 | NP_742015.1 | 2 | 193 | ||
| XM_006247874.1 | XP_006247936.1 | 2 | 386 | |||
| XM_006247875.1 | XP_006247937.1 | 2 | 386 | |||
| GenBank mRNA | AF508021.1 | - | 2 | 193 | ||
| AY327509.1 | - | 2 | 193 | |||
| AY327510.1 | - | 2 | 193 | |||

Dus1l
Genomic Map