siRNA Details
| Gene Symbol: Ifi47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172019.2 | NP_742016.2 | 2 | 815 | ||
| XM_006246175.2 | XP_006246237.1 | 2 | 1847 | |||
| XM_008767662.1 | XP_008765884.1 | 2 | 1035 | |||
| GenBank mRNA | AF508023.1 | - | 2 | 833 | ||
| BC066660.1 | - | 2 | 786 | 5623225 | ||

Ifi47
Genomic Map