siRNA Details
| Gene Symbol: Plekhb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172033.2 | NP_742030.1 | 8 | 1136 | ||
| XM_006229787.2 | XP_006229849.1 | 9 | 1374 | |||
| XM_006229788.2 | XP_006229850.1 | 8 | 1535 | |||
| XM_006229789.2 | XP_006229851.1 | 9 | 1445 | |||
| XM_006229791.2 | XP_006229853.1 | 7 | 1902 | |||
| XM_008759795.1 | XP_008758017.1 | 7 | 1426 | |||
| GenBank mRNA | AF081582.1 | - | 8 | 1136 | ||
| AF118562.1 | - | 8 | 1106 | |||
| BC078704.1 | - | 7 | 1061 | 7124581 | ||

Plekhb1
Genomic Map