siRNA Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172062.2 | NP_742059.2 | 8 | 1191 | ||
| XM_006256424.2 | XP_006256486.1 | 9 | 1276 | |||
| XM_006256425.2 | XP_006256487.1 | 8 | 1185 | |||
| GenBank mRNA | BC078703.1 | - | 8 | 1191 | 7115543 | |
| X78949.1 | - | 8 | 1025 | |||

P4ha1
Genomic Map