siRNA Details
Gene Symbol: Has1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172323.1 | NP_758826.1 | 3 | 945 | ||
GenBank mRNA | AB097568.1 | - | 3 | 945 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Has1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172323.1 | NP_758826.1 | 3 | 945 | ||
GenBank mRNA | AB097568.1 | - | 3 | 945 |