siRNA Details
| Gene Symbol: Zc3hav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173045.2 | NP_766633.2 | 4 | 1549 | ||
| XM_006236295.2 | XP_006236357.1 | 4 | 1610 | |||
| GenBank mRNA | AF521008.1 | - | 4 | 1233 | ||
| BC131842.1 | - | 4 | 1549 | 8372019 | ||
| FQ230840.1 | - | 4 | 1572 | |||

Zc3hav1
Genomic Map