siRNA Details
| Gene Symbol: Sgpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173116.1 | NP_775139.1 | 10, 11 | 1046 | ||
| XM_008772870.1 | XP_008771092.1 | 10, 11 | 985 | |||
| XM_008772871.1 | XP_008771093.1 | 10, 11 | 995 | |||
| XM_008772872.1 | XP_008771094.1 | 9, 10 | 966 | |||
| XM_008772873.1 | XP_008771095.1 | 10, 11 | 1053 | |||
| GenBank mRNA | AJ512838.1 | - | 10, 11 | 1046 | ||

Sgpl1
Genomic Map