siRNA Details
| Gene Symbol: Olr1278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173129.1 | NP_775152.1 | 2 | 1224 | ||
| GenBank mRNA | AF010293.1 | - | 2 | 1224 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1278| Gene Symbol: Olr1278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173129.1 | NP_775152.1 | 2 | 1224 | ||
| GenBank mRNA | AF010293.1 | - | 2 | 1224 | ||