siRNA Details
| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173314.2 | NP_775436.2 | 3 | 257 | ||
| XM_006232101.2 | XP_006232163.1 | 3 | 493 | |||
| GenBank mRNA | BC161833.1 | - | 3 | 257 | ||
| U67083.1 | - | 245 | ||||
| Gene Symbol: LOC680200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139491.1 | NP_001132963.1 | 3 | 287 | ||
| GenBank mRNA | BC167062.1 | - | 3 | 287 | 9024521 | |

Zfp455
Genomic Map