siRNA Details
Gene Symbol: Rsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232102.2 | XP_006232164.1 | 4 | 755 | ||
XM_006232103.2 | XP_006232165.1 | 4 | 1024 | |||
XM_006232104.2 | XP_006232166.1 | 4 | 479 | |||
XM_006232105.2 | XP_006232167.1 | 4 | 779 | |||
XM_006232106.2 | XP_006232168.1 | 4 | 487 | |||
XM_008760847.1 | XP_008759069.1 | 4 | 1067 |
Gene Symbol: Zfp455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173314.2 | NP_775436.2 | 4 | 524 | ||
XM_006232101.2 | XP_006232163.1 | 4 | 760 | |||
GenBank mRNA | U67083.1 | - | 512 |
