siRNA Details
Gene Symbol: Vom2r40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173317.1 | NP_775439.1 | 1284 | |||
GenBank mRNA | AF016180.1 | - | 1284 |
Gene Symbol: LOC679574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759645.1 | XP_008757867.1 | 3 | 644 |
Gene Symbol: Vom2r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099511.1 | NP_001092981.1 | 3 | 662 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
