siRNA Details
Gene Symbol: Zfp689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173330.1 | NP_775452.1 | 3 | 601 | ||
GenBank mRNA | AF277902.1 | - | 3 | 601 | ||
BC168646.1 | - | 3 | 609 | 9098465 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173330.1 | NP_775452.1 | 3 | 601 | ||
GenBank mRNA | AF277902.1 | - | 3 | 601 | ||
BC168646.1 | - | 3 | 609 | 9098465 |