siRNA Details
| Gene Symbol: Sgtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181629.3 | NP_853660.1 | 5 | 439 | ||
| XM_006231882.2 | XP_006231944.1 | 5 | 605 | |||
| XM_006231883.2 | XP_006231945.1 | 5 | 580 | |||
| GenBank mRNA | AF368280.1 | - | 5 | 440 | ||
| FQ211527.1 | - | 5 | 483 | |||
| FQ212077.1 | - | 5 | 607 | |||
| FQ212215.1 | - | 5 | 582 | |||

Sgtb
Genomic Map