siRNA Details
| Gene Symbol: Isl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017339.3 | NP_059035.3 | 2 | 177 | ||
| GenBank mRNA | AY557632.1 | - | 2 | 172 | ||
| S69329.1 | - | 2 | 177 | |||
| X53258.1 | - | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isl1| Gene Symbol: Isl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017339.3 | NP_059035.3 | 2 | 177 | ||
| GenBank mRNA | AY557632.1 | - | 2 | 172 | ||
| S69329.1 | - | 2 | 177 | |||
| X53258.1 | - | 2 | 280 | |||