siRNA Details
| Gene Symbol: Tp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127339.1 | NP_001120811.1 | 13 | 2406 | ||
| NM_001127342.1 | NP_001120814.1 | 12 | 2071 | |||
| NM_001127343.1 | NP_001120815.1 | 11 | 1977 | |||
| NM_019221.3 | NP_062094.1 | 14 | 2500 | |||
| GenBank mRNA | Y10258.2 | - | 14 | 2500 | ||

Tp63
Genomic Map