siRNA Details
Gene Symbol: Synpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021695.2 | NP_067727.2 | 2 | 653 | ||
GenBank mRNA | AB013130.1 | - | 1 | 179 | ||
AM980944.1 | - | 123 | ||||
CB794090.1 | - | 1 | 395 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Synpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021695.2 | NP_067727.2 | 2 | 653 | ||
GenBank mRNA | AB013130.1 | - | 1 | 179 | ||
AM980944.1 | - | 123 | ||||
CB794090.1 | - | 1 | 395 |