siRNA Details
Gene Symbol: Pgrmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021766.1 | NP_068534.1 | 2, 3 | 592 | ||
GenBank mRNA | AF163321.1 | - | 2, 3 | 550 | ||
AF465623.1 | - | 2, 3 | 498 | |||
AJ005837.1 | - | 2, 3 | 564 | |||
BC062073.1 | - | 2, 3 | 553 | 5599449 | ||
U63315.1 | - | 2, 3 | 592 |
