siRNA Details
| Gene Symbol: Vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021847.3 | NP_068619.1 | 5 | 497 | ||
| XM_006235706.2 | XP_006235768.2 | 5 | 671 | |||
| GenBank mRNA | AF086631.1 | - | 5 | 497 | ||
| BC065576.1 | - | 5 | 682 | 5598396 | ||

Vapb
Genomic Map