siRNA Details
| Gene Symbol: Matk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021859.2 | NP_068631.1 | 8, 9 | 817 | ||
| XM_006240989.2 | XP_006241051.1 | 8, 9 | 1183 | |||
| XM_006240990.2 | XP_006241052.1 | 8, 9 | 843 | |||
| XM_008765128.1 | XP_008763350.1 | 8, 9 | 1147 | |||
| GenBank mRNA | BC087726.1 | - | 8, 9 | 817 | 7320205 | |

Matk
Genomic Map