siRNA Details
| Gene Symbol: Gtf2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022208.1 | NP_071544.1 | 3 | 317 | ||
| XM_006240410.2 | XP_006240472.1 | 3 | 592 | |||
| GenBank mRNA | AF000943.1 | - | 3 | 317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf2a1| Gene Symbol: Gtf2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022208.1 | NP_071544.1 | 3 | 317 | ||
| XM_006240410.2 | XP_006240472.1 | 3 | 592 | |||
| GenBank mRNA | AF000943.1 | - | 3 | 317 | ||