siRNA Details
| Gene Symbol: Lgmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022226.2 | NP_071562.2 | 2 | 277 | ||
| GenBank mRNA | AB032766.1 | - | 2 | 272 | ||
| AF154349.1 | - | 2 | 277 | |||
| BC087708.1 | - | 314 | 7315308 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgmn| Gene Symbol: Lgmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022226.2 | NP_071562.2 | 2 | 277 | ||
| GenBank mRNA | AB032766.1 | - | 2 | 272 | ||
| AF154349.1 | - | 2 | 277 | |||
| BC087708.1 | - | 314 | 7315308 | |||