siRNA Details
| Gene Symbol: Nmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022239.2 | NP_071575.1 | 2 | 268 | ||
| XM_006250872.2 | XP_006250934.1 | 2 | 1088 | |||
| GenBank mRNA | AJ510132.1 | - | 1 | 27 | ||
| M94555.1 | - | 2 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmu| Gene Symbol: Nmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022239.2 | NP_071575.1 | 2 | 268 | ||
| XM_006250872.2 | XP_006250934.1 | 2 | 1088 | |||
| GenBank mRNA | AJ510132.1 | - | 1 | 27 | ||
| M94555.1 | - | 2 | 268 | |||