siRNA Details
| Gene Symbol: Pygl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022268.1 | NP_071604.1 | 2, 3 | 357 | ||
| GenBank mRNA | X63515.1 | - | 2, 3 | 357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pygl| Gene Symbol: Pygl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022268.1 | NP_071604.1 | 2, 3 | 357 | ||
| GenBank mRNA | X63515.1 | - | 2, 3 | 357 | ||