siRNA Details
| Gene Symbol: Nmur2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022275.3 | NP_071611.3 | 1 | 730 | ||
| GenBank mRNA | AB041229.1 | - | 1 | 730 | ||
| AF242875.1 | - | 1 | 604 | |||
More Information
Other Species Targeted
Set Membership: |
|

Nmur2
Genomic Map