siRNA Details
| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022278.1 | NP_071614.1 | 3 | 446 | ||
| GenBank mRNA | AF167981.3 | - | 3 | 446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glrx| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022278.1 | NP_071614.1 | 3 | 446 | ||
| GenBank mRNA | AF167981.3 | - | 3 | 446 | ||