siRNA Details
| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022278.1 | NP_071614.1 | 3 | 1216 | ||
| GenBank mRNA | AF167981.3 | - | 3 | 1216 | ||
| BC061555.1 | - | 3 | 1245 | 6889009 | ||
| FQ224532.1 | - | 3 | 1252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glrx| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022278.1 | NP_071614.1 | 3 | 1216 | ||
| GenBank mRNA | AF167981.3 | - | 3 | 1216 | ||
| BC061555.1 | - | 3 | 1245 | 6889009 | ||
| FQ224532.1 | - | 3 | 1252 | |||