siRNA Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022279.1 | NP_071615.1 | 14 | 1652 | ||
| GenBank mRNA | AF218574.1 | - | 14 | 1652 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mre11a| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022279.1 | NP_071615.1 | 14 | 1652 | ||
| GenBank mRNA | AF218574.1 | - | 14 | 1652 | ||